Order Entry
Northern Ireland
ContactUsLinkComponent
tert-Butyl hydroperoxide 70% in water
Flammable
Corrosive
Toxic
Health
Environment
tert-Butyl hydroperoxide 70% in water
Certificates
undefined
Flammable
Corrosive
Toxic
Health
Environment
tert-Butyl hydroperoxide 70% in water

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance:
    Clear solution
  • Titration Iodometric:
    68.5 to 71.0 %
  • Color scale:
    ≤20 APHA
  • Titration with HClO4:
    ≥97.5 % (on dry substance)

Specification Test Results

Related Information

Frequently Bought Together