
3-Methylthiophene-2-carboxaldehyde tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5834-16-2
:
Formula:C₆H₆OS
MDL Number:MFCD00005430
Boiling Pt:215 °C
Density:1,170
Flash Pt:82 °C (179 °F)
:126.18 g/mol
EINECS:227-418-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together