Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
 
Arsenazo III ≥79% for spectrophotometric determination of Th, Zr, U, Cd, Zn, Ca, Fluka™
 
 
 
Arsenazo III ≥79% for spectrophotometric determination of Th, Zr, U, Cd, Zn, Ca, Fluka™

 

  • Assay (HPLC):
    Min. 79 %
  • Colour of the solution:
    complying
  • Loss on drying (110°C):
    Max. 10 %
  • Sulfated ash:
    Max. 1 %
  • Calcium (Ca):
    Max. 500 ppm
  • Cadmium (Cd):
    Max. 50 ppm
  • Chromium (Cr):
    Max. 200 ppm
  • Magnesium (Mg):
    Max. 50 ppm
  • Molybdenum (Mo):
    Max. 50 ppm
  • Zinc (Zn):
    Max. 50 ppm
  • Absorbtion (1%/1cm):
    Min. 300
  • Absorption maximum:
    535 - 543 nm

 

 

Frequently Bought Together