


Lithium tetrachlorocuprate 0.1 M in THF
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15489-27-7
:
Formula:Cl₄CuLi₂
MDL Number:MFCD00011081
:219.24 g/mol
Density:0,910
UN:2056
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together