Order Entry
Northern Ireland
ContactUsLinkComponent
1,3-Phenylenediacetic acid 97%
Irritant
1,3-Phenylenediacetic acid 97%
Catalog # ACRO130500010
CAS Number:  
undefined
Irritant
1,3-Phenylenediacetic acid 97%
Catalog # ACRO130500010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    171 to 179 °C
  • Titration with NaOH:
    96.0 to 104.0 %

Specification Test Results

About this item

 :
Melting Pt:173…177 °C
 :194.19 g/mol
EINECS:243-332-3
 :GHS07
Storage Temperature:Ambient
CAS Number:19806-17-8
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00004341