Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulfathiazole
Sulfathiazole
Sulfathiazole
MOLE55446644-100G
:
Molekula
:
Sulfathiazole
MOLE55446644-100G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
255.32 g/mol
:
200-771-5
:
72-14-0
:
C₉H₉N₃O₂S₂
:
MFCD00005319