Order Entry
Northern Ireland
ContactUsLinkComponent
Procaine hydrochloride
Procaine hydrochloride
Catalog # PROOICRS0398
CAS Number:  
Procaine hydrochloride
Catalog # PROOICRS0398
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :272.77 g/mol
EINECS:200-077-2
UN:2811
ADR:6.1,III
CAS Number:51-05-8
Formula:C₁₃H₂₁ClN₂O₂
MDL Number:MFCD00013000