Procaine hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51-05-8
:
Formula:C₁₃H₂₁ClN₂O₂
MDL Number:MFCD00013000
:272.77 g/mol
Melting Pt:154…158 °C
Storage Temperature:Ambient
EINECS:200-077-2
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Colorless to white to almost white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:154 to 158 °C
- Loss on drying:≤0.5 % (1 g, 105 °C)
- Sulfated ash:≤0.1 %
- pH:5.0 to 6.5 (2 % w/v in water)
- Titration with NaOH:98.5 to 101.5 %
Specification Test Results
Related Information
Frequently Bought Together