Order Entry
Northern Ireland
ContactUsLinkComponent
 
Glutardialdehyde bis(sodium hydrogen sulphite) for synthesis, Sigma-Aldrich®
  8.04240.0250
 :  Merck
 :  
Glutardialdehyde bis(sodium hydrogen sulphite) for synthesis, Sigma-Aldrich®
  8.04240.0250
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Solubility:
    285 g/l (20 °C)
  • Molar Mass:
    308.22 g/mol
  • pH:
    3 - 5 (100 g/l, H2O, 20 °C)
  • Synonyms:
    Pentane-1,5-dial-bis(sodium bisulfit)
  • Hill Formula:
    C5H10Na2O8S2
  • Chemical Formula:
    CH2[CH2CH(OH)SO3Na]2

 

 

 :
 :308.24 g/mol
 :M-Clarity™ quality level = MQ200
 :231-043-5
 :7420-89-5
 :C₅H₁₀Na₂O₈S₂