

Ethylene glycolbismercaptoacetate for synthesis, Sigma-Aldrich®
Catalog # 8.14145.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Solubility:20 g/l (20 °C)
- Molar Mass:210.27 g/mol
- Boiling Point:137 - 139 °C (2 hPa)
- Density:1.32 g/cm3 (20 °C)
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specification Test Results
About this item
:
:210.28 g/mol
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:204-653-4
UN:3082
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:123-81-9
Formula:C₆H₁₀O₄S₂
MDL Number:MFCD00004875