Order Entry
Northern Ireland
ContactUsLinkComponent
(±)-Mandelonitrile for synthesis, Sigma-Aldrich®
Toxic
(±)-Mandelonitrile for synthesis, Sigma-Aldrich®
Catalog # 8.19016.0100
Supplier:  Merck
CAS Number:  
Toxic
(±)-Mandelonitrile for synthesis, Sigma-Aldrich®
Catalog # 8.19016.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

Specification Test Results

  • Solubility:
     - 1 g/l (20 °C)
  • Melting Point:
    -10 °C
  • Molar Mass:
    133.15 g/mol
  • Boiling Point:
    170 °C (1013 hPa)
  • Flash Point:
    86 °C
  • Density:
    1.12 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :133.15
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
EINECS:208-532-7
UN:3276
 :GHS06
Storage Temperature:Ambient
CAS Number:532-28-5
Formula:C₈H₇NO
MDL Number:MFCD00004487