
2-Amino-5,6-dimethyl-4-hydroxypyrimidine ≥97%
Catalog # 43432.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:139.16 g/mol
Melting Pt:316…318 °C (dec.)
EINECS:223-611-6
:GHS07
Storage Temperature:Ambient
CAS Number:3977-23-9
Formula:C₆H₉N₃O
MDL Number:MFCD00023258