Order Entry
Northern Ireland
ContactUsLinkComponent
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) for synthesis, Sigma-Aldrich®
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    1670 g/l (20 °C)
  • Melting Point:
    67 - 71 °C
  • Molar Mass:
    172.25 g/mol
  • Boiling Point:
    (1013 hPa) not applicable,(decomposition)
  • Vapor Pressure:
     - 0.01 hPa (20 °C)
  • Flash Point:
    146 °C DIN 51758
  • pH:
    8.2 (20 g/l, H2O, 20 °C)
  • Ignition Point:
    260 °C DIN 51794

Specification Test Results

Related Information

Frequently Bought Together