2,5-Piperazinedione for synthesis, Sigma-Aldrich®
Catalog # 8.14160.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:142 g/l (20 °C)
- Molar Mass:114.1 g/mol
- Synonyms:Glycinanhydride, 2,5-Diketopiperazine
- Hill Formula:C4H6N2O2
Specification Test Results
About this item
:
:114.1 g/mol
:M-Clarity™ quality level = MQ200
EINECS:203-411-5
CAS Number:106-57-0
Formula:C₄H₆N₂O₂
MDL Number:MFCD00006009