Polyethylene glycol 35,000 stabilised for synthesis pH 5 - 7 (100 g/l, H₂O, 20 °C), Sigma-Aldrich®
: Merck
:25322-68-3
:
:H(OCH₂CH₂)nOH
:MFCD00081839
:35000 g/mol
:60…65 °C
:1,2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:260 °C
:Stabilised with 2-tert-butyl-4-methoxyphenol
:Ambient
:500-038-2
- Hydroxyl value:3-4
- Melting range (lower value):≥ 62 °C
- Melting range (upper value):≤ 67 °C
- Average molecular mass:28000 - 38000
- Identity (IR):passes test
Frequently Bought Together