
2-(Ethylthio)ethanol for synthesis, Sigma-Aldrich®
Catalog # 8.40111.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 ml
- Seq:2315
Specifications
Specification Test Results
- Melting Point:-100 °C
- Molar Mass:106.19 g/mol
- Boiling Point:180 - 184 °C (1013 hPa)
- Flash Point:93 °C
- Density:1.018 g/cm3 (20 °C)
Specification Test Results
About this item
:
:106.19 g/mol
:M-Clarity™ quality level = MQ200
EINECS:203-802-0
:GHS05
Storage Temperature:Ambient
CAS Number:110-77-0
Formula:C₄H₁₀OS
MDL Number:MFCD00002909