£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
3-Phenyl-1H-pyrazole
3-Phenyl-1H-pyrazole for synthesis, Sigma-Aldrich®
3-Phenyl-1H-pyrazole for synthesis, Sigma-Aldrich®
8.43855.0005
:
Merck
:
3-Phenyl-1H-pyrazole for synthesis, Sigma-Aldrich®
8.43855.0005
:
Merck
:
Pack type:
Glass bottle
Pk:
5 g
Seq:
3173
:
:
144.18 g/mol
:
M-Clarity™ quality level = MQ200
:
GHS07
:
Ambient
:
2458-26-6
:
C₉H₈N₂
:
MFCD00159654