
Lithium salicylate, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:552-38-5
:
Formula:C₇H₅LiO₃
MDL Number:MFCD00045813
:144.06 g/mol
Melting Pt: Dec.
EINECS:209-011-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together