Order Entry
Northern Ireland
ContactUsLinkComponent
1-Bromo-3-methoxypropane for synthesis, Sigma-Aldrich®
1-Bromo-3-methoxypropane for synthesis, Sigma-Aldrich®
Catalog # 8.43834.0025
Supplier:  Merck
CAS Number:  
1-Bromo-3-methoxypropane for synthesis, Sigma-Aldrich®
Catalog # 8.43834.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Molar Mass:
    153.02 g/mol
  • Boiling Point:
    121 °C (1013 hPa)
  • Vapor Pressure:
    6.6 hPa (20 °C)
  • Flash Point:
    43 °C
  • Density:
    1.35 g/cm3
  • pH:
    6 - 7 (H2O)
  • Hill Formula:
    C4H9BrO

Specification Test Results

About this item

 :
 :153.02 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ200
UN:1993
 :GHS02
 :GHS06
Storage Temperature:Ambient
CAS Number:36865-41-5
Formula:C₄H₉BrO
MDL Number:MFCD02258473