Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Goggles
VWR®, Replacement Lens for VWR® Autoclavable Cleanroom Goggles
VWR®, Replacement Lens for VWR® Autoclavable Cleanroom Goggles
111-0599
:
VWR Collection
VWR®, Replacement Lens for VWR® Autoclavable Cleanroom Goggles
111-0599
:
VWR Collection
Colour:
Clear
Description:
Replacement lens for autoclavable cleanroom goggles
Pk:
10