Tubing, FEP, BOLA
Supplier: Bohlender
Certificates
About this item
PFA, transparent.
- Gastight tubing
- Almost universal chemical resistance
- Good sliding and dielectric characteristics
- Can be sterilized in autoclaves
- Resistant to irradiation and weather
Applications include perfect tubing for aggressive and pure liquids or gases. It has good bending radius and burst pressure.
Temperature resistance: −200 to +260 °C.
Frequently Bought Together