ContactUsLinkComponent
Test strips, chlorine (free), Reflectoquant®, Supelco®
Test strips, chlorine (free), Reflectoquant®, Supelco®
  1.16896.0001
 :  Merck
 :  
undefined
Test strips, chlorine (free), Reflectoquant®, Supelco®
  1.16896.0001
 :  Merck
 :  
  • Chemical method:
    Redox reaction
  • Pk:
    50 Tests
  • Description:
    Chlorine test (free chlorine)
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ100
  • Type:
    RQ
  • Range:
    0,5 - 10,0 mg/l Cl₂
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly