ContactUsLinkComponent
Burette and test tube clamps, with bosshead
Burette and test tube clamps, with bosshead
Catalog # BOCH5631
Supplier:  BOCHEM
undefined
Burette and test tube clamps, with bosshead
Catalog # BOCH5631
Supplier:  BOCHEM
Specifications
  • For:
    Burettes and test tubes
  • Spanning width:
    7 - 25 mm
  • Pk:
    1
Specifications
About this item
These chromium plated clamps with bosshead (13,5 mm) and PVC coated jaws are designed for holding burettes and test tubes (Ø 7 - 25 mm).