1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23978-55-4
:
Formula:C₁₂H₂₆N₂O₄
MDL Number:MFCD00005112
:262.35 g/mol
Melting Pt:111…115 °C
Storage Temperature:Refrigerator
EINECS:245-965-0
UN:3259
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Off-white Glistening crystalline powder
- Infrared spectrum:Conforms
- Melting point:111 to 115 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together