
1-Hydroxycyclohexyl phenyl ketone ≥98%
Catalog # L11377.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:947-19-3
:
Formula:C₁₃H₁₆O₂
MDL Number:MFCD00059561
Boiling Pt:>225 °C
Melting Pt:46…49 °C
Density:1,170
Flash Pt:>150 °C (302 °F)
:204.27 g/mol
EINECS:213-426-9
:GHS07
Storage Temperature:Ambient