2,2-Difluoropropionic acid ≥97%
Catalog # H26030.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2,2-Difluoropropanoic acid
CAS Number:373-96-6
:
Formula:C₃H₄F₂O₂
MDL Number:MFCD03093764
:110.06 g/mol
Flash Pt:66 °C (150 °F)
UN:3261
:GHS05
Boiling Pt:142 °C
Melting Pt:36…44 °C
Storage Temperature:Ambient
ADR:8 II