
Isopentyl propionate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:105-68-0
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00048611
:144.21 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:203-322-1
UN:3272
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Melting Point:-73 °C
- Molar Mass:144.21 g/mol
- Boiling Point:160 °C
- Flash Point:46 °C
- Explosion Limit:1 %(V)
- Density:0.869 g/cm3 (20 °C)
- Ignition Point:370 °C
Specification Test Results
Frequently Bought Together