
2-Ethylhexyl phosphate (mono and di ester mixture) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12645-31-7
:
MDL Number:MFCD00135884
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:235-741-0
UN:3265
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble
- Boiling Point:180 °C (1013 hPa) decomposes
- Flash Point:100 °C
- Density:1.02 g/cm3 (20 °C)
- pH:2.3 (10 g/l, H2O, 20 °C) (as an emulsion)
Specification Test Results
Frequently Bought Together