Order Entry
Northern Ireland
ContactUsLinkComponent
N-(4-aminophenyl)aniline hydrochloride for synthesis, Sigma-Aldrich®
Irritant
N-(4-aminophenyl)aniline hydrochloride for synthesis, Sigma-Aldrich®
Catalog # 8.00405.0100
Supplier:  Merck
CAS Number:  
Irritant
N-(4-aminophenyl)aniline hydrochloride for synthesis, Sigma-Aldrich®
Catalog # 8.00405.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    258 - 261 °C
  • Molar Mass:
    220.7 g/mol

Specification Test Results

About this item

 :
 :220.7 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:260-173-5
 :GHS07
Storage Temperature:Ambient
CAS Number:56426-15-4
Formula:C₁₂H₁₃ClN₂