4-Tritylphenol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:978-86-9
:
Formula:C₂₅H₂₀O
MDL Number:MFCD00002364
:336.43 g/mol
Melting Pt:276…286 °C
EINECS:213-557-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together