Order Entry
Northern Ireland
ContactUsLinkComponent
3-Methylthiophene 99+%
Irritant
Flammable
3-Methylthiophene 99+%
Certificates
undefined
Irritant
Flammable
3-Methylthiophene 99+%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5180 to 1.5210 (20°C, 589 nm)
  • GC:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together