Order Entry
Northern Ireland
ContactUsLinkComponent
2,6-Lutidine for synthesis, Sigma-Aldrich®
Flammable
Irritant
2,6-Lutidine for synthesis, Sigma-Aldrich®
Catalog # 8.05650.0250
Supplier:  Merck
CAS Number:  
Flammable
Irritant
2,6-Lutidine for synthesis, Sigma-Aldrich®
Catalog # 8.05650.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -6 °C
  • Molar Mass:
    107.15 g/mol
  • Boiling Point:
    144 °C (1013 hPa)
  • Vapor Pressure:
    5.5 hPa (20 °C)
  • Flash Point:
    33 °C
  • Refractive Index:
    1.4976 (20 °C, 589 nm)
  • Density:
    0.923 g/cm3 (20 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

About this item

 :
 :107.16 g/mol
Merck Index:13,05636
Boiling Pt:143,7 °C (1013 hPa)
Melting Pt:–6 °C
Density:0,9252 g/cm³ (20 °C)
Flash Pt:33 °C
EINECS:203-587-3
UN:1993
ADR:3,III
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:108-48-5
Formula:C₇H₉N
MDL Number:MFCD00006345