Order Entry
Northern Ireland
ContactUsLinkComponent
4-Picoline for synthesis, Sigma-Aldrich®
Flammable
Toxic
4-Picoline for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Toxic
4-Picoline for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1000 g/l (25 °C)
  • Melting Point:
    3 °C
  • Molar Mass:
    93.13 g/mol
  • Boiling Point:
    145 °C (1013 hPa)
  • Vapor Pressure:
    5.8 hPa (20 °C)
  • Flash Point:
    40 °C
  • Refractive Index:
    1.5049 (20 °C, 589 nm)
  • Explosion Limit:
    1.3 - 8.7 %(V)
  • Density:
    0.95 g/cm3 (20 °C)
  • Ignition Point:
    500 °C

Specification Test Results

Related Information

Frequently Bought Together