


2,6-Diaminopyridine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:141-86-6
:
Formula:C₅H₇N₃
MDL Number:MFCD00006329
:109.13 g/mol
Boiling Pt:288 °C
Melting Pt:118…121 °C
Flash Pt:155 °C (311 °F)
EINECS:205-507-2
UN:2671
ADR:6.1,II
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together