
2-Chloro-5-nitropyridine ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4548-45-2
:
Formula:C₅H₃ClN₂O₂
MDL Number:MFCD00006240
:158.54 g/mol
Melting Pt:104…109 °C
Flash Pt:158 °C (316 °F)
EINECS:224-908-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together