


Tetrahydrofuran, Uvasol® for spectroscopy, Supelco®
: Merck
:109-99-9
:
:C₄H₈O
:MFCD00005356
:72.11 g/mol
:12,09351
:66 °C (1013 hPa)
:–108,5 °C
:0,888 g/cm³ (20 °C)
:Ambient
:203-726-8
:2056
:3,II
:M-Clarity™ quality level = MQ100
:–21,5 °C
:05-2114615188-48
:GHS02
:GHS08
:GHS07
- Solubility:(20 °C) soluble
- Melting Point:-108.5 °C
- Molar Mass:72.11 g/mol
- Boiling Point:65 - 66 °C (1013 hPa)
- Vapor Pressure:173 hPa (20 °C)
- Flash Point:-20 °C
- Explosion Limit:1.5 - 12.4 %(V)
- Density:0.89 g/cm³ (20 °C)
- pH:7 - 8 (200 g/L, H₂O, 20 °C)
- Ignition Point:215 °C
- Saturation Concentration:557 g/m³ (20 °C) Air
- Evaporation Number:2.3
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Frequently Bought Together