ContactUsLinkComponent
Industrial gloves, Polytril™ Air ³/₄
Industrial gloves, Polytril™ Air ³/₄
Supplier:  HONEYWELL SAFETY
Certificates
undefined
Industrial gloves, Polytril™ Air ³/₄
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • 112-1533
  • 112-1536
  • 112-1535
  • 112-1534
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Finish:
  • Palm and fingers coated
  • Palm and fingers coated
  • Palm and fingers coated
  • Palm and fingers coated
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)