ContactUsLinkComponent
General purpose gloves, Perfect Poly® grey
General purpose gloves, Perfect Poly® grey
Supplier:  HONEYWELL SAFETY
Certificates
undefined
General purpose gloves, Perfect Poly® grey
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • DALL2400250/11
  • DALL2400250/10
  • DALL2400250/9
  • DALL2400250/8
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Finish:
  • Coating on palm and fingertips
  • Coating on palm and fingertips
  • Coating on palm and fingertips
  • Coating on palm and fingertips
  • Gauge:
  • 15
  • 15
  • 15
  • 15
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)