Order Entry
Northern Ireland
ContactUsLinkComponent
2-(Bromomethyl)naphthalene 96%
Corrosive
2-(Bromomethyl)naphthalene 96%
Certificates
undefined
Corrosive
2-(Bromomethyl)naphthalene 96%

Specification Test Results

  • Appearance:
    White to light yellow to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    52 to 57 °C
  • GC:
    ≥95 %

Specification Test Results

Related Information

Frequently Bought Together