
2-Chloronaphthalene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:91-58-7
:
Formula:C₁₀H₇Cl
MDL Number:MFCD00035731
:162.62 g/mol
Merck Index:13,02169
Boiling Pt:256 °C (760 mmHg)
Melting Pt:57…60 °C
Density:1,2656 g/cm³ (16 °C)
Flash Pt:125 °C
EINECS:202-079-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:57 to 60 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together