
2-Bromonaphthalene 99%
:580-13-2
:
:C₁₀H₇Br
:MFCD00004051
:13,01411
:281…282 °C (760 mmHg)
:53…57 °C
:1,605 g/cm³ (0 °C)
:>110 °C
:207.07 g/mol
:209-452-5
:GHS07
:Ambient
- Appearance:White to light yellow to light beige Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:53 to 57 °C
- GC:≥98.5 %
Frequently Bought Together