

Nickel(II) telluride ≥99.9% (metals basis excluding Co) (max. 0.1-1% Co)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12142-88-0
:
Formula:NiTe
MDL Number:MFCD00016258
:186.29 g/mol
Storage Temperature:Ambient
EINECS:235-260-6
UN:3077
ADR:9,III
:GHS08
:GHS07
Related Information
Frequently Bought Together