
Chromium(III) oxide ≥99.8% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41056.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:151.99 g/mol
Merck Index:13,02247
Boiling Pt:3000 °C
Melting Pt:2435 °C
Density:5,22
Storage Temperature:Ambient
EINECS:215-160-9
:GHS07
CAS Number:1308-38-9
Formula:Cr₂O₃
MDL Number:MFCD00010949