Zirconium tetrafluoride ≥99.9% (metals basis)
Catalog # 39489.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:167.22 g/mol
Merck Index:14,10175
Melting Pt:600 °C (subl.)
Density:4,43
Storage Temperature:Ambient
EINECS:232-018-1
UN:3260
ADR:8,II
:GHS05
CAS Number:7783-64-4
Formula:F₄Zr
MDL Number:MFCD00011307