
Praseodymium(III,IV) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12037-29-5
:
Formula:O₁₁Pr₆
MDL Number:MFCD00011178
:1021.44 g/mol
Melting Pt:2500 °C
EINECS:234-857-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together