

Praseodymium trifluoride, anhydrous ≥99.9895% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13709-46-1
:
Formula:F₃Pr
MDL Number:MFCD00016287
Melting Pt:1395 °C
Density:6,2
Storage Temperature:Ambient
EINECS:237-254-9
UN:3288
ADR:6.1,III
:197.9 g/mol
:GHS06
:GHS07
Related Information
Frequently Bought Together