
Lanthanum(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13709-38-1
:
Formula:F₃La
MDL Number:MFCD00011073
:195.9 g/mol
Boiling Pt:∼2327 °C (1013 hPa)
Melting Pt:1493 °C
Density:5,936
Storage Temperature:Ambient
EINECS:237-252-8
UN:3288
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together