£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
4-Bromoisoquinoline
4-Bromoisoquinoline
4-Bromoisoquinoline
MOLE17966472-25G
:
Molekula
:
4-Bromoisoquinoline
MOLE17966472-25G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
208.06 g/mol
:
280 °C (1013 hPa)
:
42…42,5 °C
:
216-244-8
:
GHS07
:
Refrigerator
:
1532-97-4
:
C₉H₆BrN
:
MFCD00006904