


Diphenyl diselenide for synthesis, Sigma-Aldrich®
Catalog # 8.18061.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:58 - 61 °C
- Molar Mass:312.13 g/mol
Specification Test Results
About this item
:
Boiling Pt:377 °C (1013 hPa)
Melting Pt:58…61 °C
Density:1,32 g/cm³ (20 °C)
UN:3283
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:312.13 g/mol
EINECS:216-780-2
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:1666-13-3
Formula:C₁₂H₁₀Se₂
MDL Number:MFCD00003001