


Selenium tetrabromide ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-65-3
:
Formula:Br₄Se
MDL Number:MFCD00049766
:398.58 g/mol
Merck Index:14,8437
Melting Pt:70…80 °C (dec.)
Density:4,029 g/cm³ (–78 °C)
EINECS:232-181-9
UN:2923
ADR:8,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together